Orders LinkContactUsLinkComponent
Ward's® Labradorite (Cleavage)
Supplier:   Avantor
Ward's® Labradorite (Cleavage)
Supplier:  Avantor
Certificates
Ward's® Labradorite (Cleavage)
Ward's® Labradorite (Cleavage)
4 Options Available Below
Product Details & Documents
Feldspar group; good cleavages of pure soda lime feldspar; most show twinning and labradorescence.
  • Collected in Quebec
  • Multiple sizes and pack options to fit your specific needs
  • Cut by Ward's geologists to guarantee top market quality
  • Matching thin section available
Student specimens - Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand specimens - Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk pack - Includes 1 kilo of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25" to 2×3". Packaged in a labeled storage box for easy organization.
All specimen dimensions are considered an average size range.
Product Family Options
Specimens
Información del producto
  • Specimen Dimensions
    Availability
    Price
  • 470024-946
    Labradorite Thin Section
    Labradorite Thin Section
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $900.60
    $900.60
    Each (1UN)
       $900.60
  • 470025-658
    Labradorite (Cleavage), Student Specimens
    Labradorite (Cleavage), Student Specimens
    1×1 - 1×1"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 10
    Paquete de 10 - $296.10
    $296.10
    Paquete de 10
       $296.10
  • 470025-662
    Labradorite (Cleavage), Hand Specimen
    Labradorite (Cleavage), Hand Specimen
    2×3" - 3×4"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $376.30
    $376.30
    Each (1UN)
       $376.30
  • 470025-660
    Labradorite (Cleavage), Bulk Pack, 1 kg
    Labradorite (Cleavage), Bulk Pack, 1 kg
    Size will vary
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $495.43
    $495.43
    Each (1UN)
       $495.43