Laboratory Spectacles
Laboratory spectacles - adult, Scratch-resistant/hardcoat
Supplier: VWR International
Jump to:
Product Details & Documents
These adjustable laboratory spectacles provide optimum comfort.
- Single-piece lens with integrated side-shield
- Clear, wrap-around lenses
- Adjustable temple to fit different face profiles
- Polycarbonate lenses block 99% of UV light
With pliable, durable frames, these comfortable and attractive glasses deliver high-impact specifications.
These lenses provide a wide field of vision and excellent protection in laboratory settings. Colors may vary.
Certifications: Meet ANSI Z87+ 1-2010 and CSA Z94.3-2003 specifications.
Documents
Certificates
Buscar por número de loteEspecificaciones
- Description:Laboratory spectacles - adult
- Lens Coating:Scratch-resistant/hardcoat