Orders LinkContactUsLinkComponent
Laboratory Safety Mat
Laboratory Safety Mat, Red, ⅛", 9 1/2 × 9 1/2"
  470016-916
 :  MATRED
Laboratory Safety Mat
  470016-916
Laboratory Safety Mat
Laboratory Safety Mat
Laboratory Safety Mat, Red, ⅛", 9 1/2 × 9 1/2"
$1,046.81
Each (1UN)
Stabilize glassware on orbital or rocking shakers and eliminate slippery conditions on benchtop surfaces with this versatile non-slip safety mat.
Its ribbed, textured surface steadily holds beakers, cylinders, and other glassware in place, even at an angle. A valuable addition to any laboratory, the silicone rubber mat can be autoclaved, cleaned with disinfectants such as bleach, easily cut to various sizes, and will withstand temperatures up to 260°C.
Size: 9 1/2" square x 1/8" thick.
  • Color:
    Red
  • Description:
    Laboratory Safety Mat
  • Thickness:
    ⅛"
  • W×L:
    9 1/2 × 9 1/2"