Orders LinkContactUsLinkComponent
Laboratory Jacks
Laboratory Jacks
Laboratory Jacks
Certificates
Laboratory Jacks
Laboratory Jacks
2 Options Available Below
Product Details & Documents
Laboratory support jack is constructed of painted aluminum and stainless steel.
  • Scissor jack design enables height adjustment with a simple turn of the knob
  • Sturdy design can support loads up to 20 kg
  • Platform height can be vertically adjusted from closed position of 2.75" up to 10.5"
  • Pre-drilled holes for attaching a support rod
Available as a set with removable support rod included.
Product Family Options
Información del producto
  • Maximum Height
    Maximum Load
    Minimum Height
    Platform Size
    Availability
    Price
  • 470001-938
    Lab Jack with removable support rod (length 18", diam. 1/2")
    Lab Jack with removable support rod (length 18", diam. 1/2")
    10.5"
    20 kg
    2.75"
    8×8"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $3,290.25
    $3,290.25
    Each (1UN)
       $3,290.25
    Supplier Number: LBJSET
  • 470199-870
    Lab Jack with removable support rod (length 18", diam. 1/2")
    Lab Jack with removable support rod (length 18", diam. 1/2")
    10.5"
    20 kg
    2.75"
    8×8"
    Discontinued
    Supplier Number: LBJSET
  • 470006-698
    Lab Jack with pre-drilled holes for attaching a support rod (not included)
    Lab Jack with pre-drilled holes for attaching a support rod (not included)
    10.5"
    20 kg
    2.75"
    4.5×5.5"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $2,784.45
    $2,784.45
    Each (1UN)
       $2,784.45
    Supplier Number: LBJK01