Orders LinkContactUsLinkComponent
Laboratory Blender/Mixer
Laboratory Blender/Mixer
155 VAC, 17.9×39.1 cm (7×14.5")
Catalog # 470006-518
Supplier:   CONAIR CORPORATION
Número de proveedor #:  700G
Laboratory Blender/Mixer
Catalog # 470006-518
Supplier:  CONAIR CORPORATION
Laboratory Blender/Mixer
Laboratory Blender/Mixer
155 VAC, 17.9×39.1 cm (7×14.5")
Price
$12,147.24
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
Heavy-duty.
  • Designed for shredding, blending, and emulsification
  • Features stainless steel blades and lubricated bearings
Especificaciones
  • Electrical:
    155 VAC
  • Diameter ×L:
    17.9×39.1 cm (7×14.5")