ContactUsLinkComponent
K’NEX Education Intro to Simple Machines: Levers and Pulleys
  470174-616
undefined
K’NEX Education Intro to Simple Machines: Levers and Pulleys
  470174-616
 :  78610
  • Descripción:
    K’NEX Intro to Simple Machines: Levers and Pulleys
  • Referencia:
    470174-616
  • Nº de proveedor:
    78610
Identify All Three Classes of Levers and Their Key Parts
After working with this set, your students will be able to discuss how levers change the direction and/or amount of force applied, identify different types of pulley systems and their key parts, and discuss how pulleys change the direction and/or amount of force applied.
Enough materials are included for one group of students to build eight different models, one at a time.
 : The set includes 178 K’NEX parts, a teacher’s guide on CD, and building instructions for each model with photos and key facts.