Kimwipes
38 cm (15"), 43 cm (17"), Box/140
Supplier: KIMBERLY-CLARK CORP (WIPERS)
Número de proveedor #: 34256
Jump to:
Product Details & Documents
Dependable, highly absorbent tissues.
- Specially designed for lab use
- Absorb up to 15 times their weight in liquid
Perfect for polishing glassware or cleaning instruments because they will not scratch or deposit lint. Supplied in handy, pop-up dispensing cartons.
Ordering information: Ordering Information: Now available in handy convenience packs. Great for classrooms!
Documents
Especificaciones
- Width:43 cm (17")
- Length:38 cm (15")
- Packaging:Box/140