Orders LinkContactUsLinkComponent
Ward's® Killing Jars
Ward's® Killing Jars
Supplier:   Avantor
Ward's® Killing Jars
Supplier:  Avantor
Certificates
Ward's® Killing Jars
Ward's® Killing Jars
1 Options Available Below
Product Details & Documents
The improved design of these jars incorporates a bottom pad that better absorbs killing agent, so that fumes are contained and insects are knocked down quickly, causing less damage to specimens.
  • Absorbent pad contains the fumes
Just one application lasts for hours, and the permanent label provides instructions for use.
Product Family Options
Información del producto
  • Availability
    Price
  • 470018-464
    WARD’S Killing Jars, 8oz. Jar
    WARD’S Killing Jars, 8oz. Jar
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $156.35
    $156.35
    Each (1UN)
       $156.35
  • 470018-466
    WARD’S Killing Jars, 16oz. Jar
    WARD’S Killing Jars, 16oz. Jar
    Discontinued