Entire Kidney Slide
Entire Kidney, Medial longitudinal section, Hematoxylin and Eosin
Catalog # 470182-922
Supplier: Avantor
Jump to:
Product Details & Documents
Observe Structures of the Kidney
- Identifying features clearly distinguishable
- Rigorous quality control standards
Rat. Through cortex, medulla, pelvis, and entrance of ureter.
Especificaciones
- Description:Entire Kidney
- Stain:Hematoxylin and Eosin
- Orientation:Medial longitudinal section