Kidney, Human Slide
Kidney, Human Slide, Section, Periodic acid Schiff reaction and Hematoxylin
Catalog # 470183-078
Supplier: Avantor
Jump to:
Product Details & Documents
Illustrates detailed kidney structure
- Identifying features clearly distinguishable
- Rigorous quality control standards
Stained for basement membrane of tubules, Bowman’s capsules, and brush borders of proximal tubules.
Especificaciones
- Description:Kidney, Human Slide
- Stain:Periodic acid Schiff reaction and Hematoxylin
- Orientation:Section