Jejunum, Human Slide
Jejunum, Human Slide, Section, Hematoxylin and Eosin
Catalog # 470183-054
Supplier: Avantor
Jump to:
Product Details & Documents
Midsection of intestine
- Identifying features clearly distinguishable
- Rigorous quality control standards
Human. Absorbs nutrients. Numerous circularly arranged folds of the mucosa and submucosa, known as plicae circulares.
Specifications
- Description:Jejunum, Human Slide
- Stain:Hematoxylin and Eosin
- Orientation:Section