Orders LinkContactUsLinkComponent
Iron Meteorite
 :   Avantor
Iron Meteorite
 :  Avantor
Iron Meteorite
Iron Meteorite
Fragments in natural condition, many showing concave forms and fusion crusts. Sizes are approximate.
  • Available in two sizes
  • Slicked in oil to prevent rusting
  • Protected in bubble wrap and a sealed bag
Product Family Options
  • Description
    Specimen Dimensions
  • Iron Meteorite
    ⅜ - ¾"
    Each (1UN)
    Each (1UN) - $295.93
    $295.93
    Each (1UN)
       $295.93
     : -
  • Iron Meteorite
    ¾ - 1"
    Each (1UN)
    Each (1UN) - $220.20
    $220.20
    Each (1UN)
       $220.20
     : -
  • Iron Meteorite
    1 - 1½"
     : TUCSON0026