ContactUsLinkComponent
Ionic Interaction Lab Activity
  470007-768
undefined
Ionic Interaction Lab Activity
  470007-768
 :  4-104
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Descripción:
    Ionic Interaction Lab Activity
  • No. of Lab Groups:
    6
  • Referencia:
    470007-768
  • Nº de proveedor:
    4-104
This kit provides an introduction to ionic interactions by giving students exposure to a large number of reactions and allowing maximum observation for a limited amount of time and effort.
  • Students Write Ionic Formulas and Equations
  • Lab Includes Follow-Up Activities
  • Test Can Be Repeated 100 Times
  • Ideal for High School Chemistry or Physical Science Studies
  • Activities Help Students Use Empirical Evidence to Draw Conclusions
It includes precipitate forming, gas forming, and non-ionized product forming reactions. The investigation may be extended to teach students a number of additional basic chemical skills such as writing ionic formulas for the products formed, writing ionic interaction equations, relating placement of elements in the periodic table to their reactivity, and developing solubility principles from empirical evidence.
This kit is ideal for high school chemistry or physical sciences studies.
 : This kit includes depression plates, toothpicks, plastic, calcium nitrate, potassium sulfate, cadmium nitrate, sodium silicate, cobalt nitrate, potassium oxalate, barium nitrate, potassium phosphate, cupric chloride, potassium chromate, potassium iodide, acetic acid, manganese nitrate, potassium hexacyanoferrate (iii) (ferricyanide), nickel nitrate, potassium hexacyanoferrate (ii) (ferrocyanide), strontium nitrate, silver nitrate, lead nitrate, ferric nitrate, zinc nitrate, chromium nitrate, aluminum nitrate , potassium bromide, potassium hydroxide, potassium chloride, potassium carbonate, and test tubes.