Introductory Histology Slide Set
Introductory Histology Slide Set, 15, Human Body
Catalog # 470177-812
Supplier: Avantor
Jump to:
Product Details & Documents
Illustrates Biological Tissues and Components
- Identifying features clearly distinguishable
- Rigorous quality control standards
Fifteen slides of basic tissue and organ systems suitable for general science and biology courses as well as an introduction to physiology or histology.
Ordering information: Set includes Ovary, mammal (cs); Parathyroid and Thyroid (sec); Testes, human (cs); Cartilage, elastic and hyaline (cs); Bone, ground (human) (cs); Muscle types, smooth striated and cardiac (ls); Spinal cord, human (cs); Cerebellum, purkinje cells (cs); Cerebrum, pyramidal cells (cs); Pig liver (cs); Fundic stomach (sec); Rabbit tongue, foliate papillae (cs); Blood, human (Wright’s stain, sm); Lymph vessels mesentery, mammal (sec); and Artery vein and nerve (cs).
Especificaciones
- Type:Human Body
- Description:Introductory Histology Slide Set
- No. of Slides:15