Introductory Histology Slide Set
Illustrates Tissue Types and Their Relation to Various Organs
- All identifying features are clearly distinguishable
- Rigorous quality control standards
- In-house experts available to answer your questions
Investigate different tissue types and organs. Slide mounting preparation varies as noted. Set of 25 slides.
: Set includes: Tissues: Squamous epithelium (sm); Areolar (wm); Cartilage (hyaline, elastic) (sect); Intramembranous ossification (sect); Bone development (cs); Giant multipolar motor neurons (sm); Pyramidal cells and their processes (sect); Nissl bodies in neurocytes (sect); Organs: Skin (bird, hairy mammal) (sect); Muscle, three types (sect); Medullated nerve (cs/ls); Spinal cord, mammal (cs); Artery, vein, and nerve (cs); Thyroid gland (sect); Submaxillary gland (sect); Esophagus-stomach junction (cs/ls); Jejunum (cs); Colon (cs); Liver (sect); Kidney, small mammal (sect); Urinary bladder, collapsed (cs); Testis, spermatogenesis (sect); Ovary (sect)
- Type:Animal
- Description:Introductory Histology Slide Set
- No. of Slides:25