Ward's® Intro to Petrography Thin Sections and Rock Set
Catalog # 470024-962
Supplier: Avantor
Specifications
- Descripción:Introduction to Petrography Thin Section and Rock Set
- No. of Slides:9
- Referencia:470024-962
- Nº de proveedor:WARD470024-962
Specifications
About this item
Set of Nine Thin Sections and Matching Rocks
- Hand Made by Wards® staff Geologists
- 9 Specially Selected Thin Sections and Matching Rock Samples
Students examine thin sections and matching 1" x 1" specimens of nine different rocks to gain a basic knowledge of systematic classification.The set contains three igneous, three sedimentary, and three metamorphic thin sections and corresponding rocks. The set also includes microscopic and macroscopic descriptions of each rock, as well as an introduction to polarized light studies.
Contents:
Igneous: diorite, gabbro, granite
Sedimentary: arkose, limestone, sandstone
Metamorphic: gneiss, phyllite, schist