ContactUsLinkComponent
Introduction And Use Of Dichotomous Keys Kit
Introduction And Use Of Dichotomous Keys Kit
Catalog # 470004-554
undefined
Introduction And Use Of Dichotomous Keys Kit
Catalog # 470004-554
Supplier Number:  51
Specifications
  • Descripción:
    Introduction And Use Of Dichotomous Keys Kit
  • No. of Lab Groups:
    30
  • Referencia:
    470004-554
  • Nº de proveedor:
    51
Specifications
About this item
This Dichotomous Keys kit enhances reading and observation skills.
  • Kit Teaches Classification Keys and Their Formats
  • Students Hone Observation and Reading Skills
  • Reusable
  • Time Required: One 50 Minute Class Period
  • Teacher's Guide Included
All dichotomous keys are developed the same way—similarities and differences in characteristics and traits are observed and recorded. Characteristics in dichotomous keys are typically given in pairs of opposing statements. The observer must make a choice as to which statement most accurately describes the specimen since statement in each pair can be true. The statement chosen determines the next pair of opposing statements. Statement pairs become increasingly more specific until the specimen is identified.
In this game-like activity, students focus on the physical attributes of an imaginary organism. They identify the specimen and draw two organisms that match the organism's description.
Ordering information: This kit contents includes a teacher's guide, student worksheets and guides, dichotomous key charts, and sets containing eight "specimen" cards.