Ward's® Internal Animal Parasites Poster
Catálogo # 470023-254
Proveedor: Avantor
Especificaciones
- Anchura:53 cm
- Descripción:Animal Parasites Poster
- Longitud:86 cm
- Referencia:470023-254
- Se ajuste:Unlaminated
- Nº de proveedor:WARD470023-254
Especificaciones
Acerca de este artículo
An Indispensable Reference For Your Invertebrate Or Agriscience Classes.
- Illustrates the life cycles of three types of parasites: flukes, tapeworms, and roundworms
Parasites can be found in almost any type of animal. The photographs and accompanying text outline their development, habitats, and mechanisms of infection.