Orders LinkContactUsLinkComponent
Insect Pinning Block
Insect Pinning Block
  470149-222
Insect Pinning Block
  470149-222
Insect Pinning Block
Insect Pinning Block
Insect Pinning Block
$62.89
Each (1UN) Case de 12 (1UN)
Traditional, 3-step wood pinning block has pin holes set at 23, 12 and 7 mm. Size: 1" x 3".
  • Description:
    Insect Pinning Block