Orders LinkContactUsLinkComponent
Increment Borers
Increment Borers
Supplier:   PECO SALES INC SE
Increment Borers
Supplier:  PECO SALES INC SE
Certificates
Increment Borers
Increment Borers
2 Options Available Below
Product Details & Documents
Used to Examine Wood Quality and Measure Growth Rates
These borers are made of the finest steel and coated with Teflon to reduce friction heat, which is an important consideration when working with growing trees. The Teflon, in combination with the three-thread style, also makes the borers easy to extract.
Product Family Options
Product Information
  • Availability
    Price
  • 470018-548
    Increment Borer, 8" Bore Length
    Increment Borer, 8" Bore Length
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $9,568.02
    $9,568.02
    Each (1UN)
       $9,568.02
    Supplier Number: 63081
  • 470018-550
    Increment Borer, 10" Bore Length
    Increment Borer, 10" Bore Length
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $10,829.11
    $10,829.11
    Each (1UN)
       $10,829.11
    Supplier Number: 63082