Orders LinkContactUsLinkComponent
Boreal Science Compound Beginner Microscopes
Boreal Science Compound Beginner Microscopes
Boreal Science Compound Beginner Microscopes
Certificates
Boreal Science Compound Beginner Microscopes
Boreal Science Compound Beginner Microscopes
2 Options Available Below
Product Details & Documents
Boreal beginner microscopes are ideal for introducing students to microscopy.
  • All-Glass-Coated Optics
  • All-Metal Construction
  • Parfocal and Parcentered
  • Objectives: 4x, 10x, and 40x
  • Boreal Lifetime Warranty
It's scaled down for younger students and is simple to use, but it also prepares students for the transition to high school instruction.
Rugged metal components are engineered for years of reliable performance. Student-proof features minimize loss of parts.
Ordering information: This product includes a manufacturer's warranty.
Product Family Options
Product Information
  • Illumination
    Availability
    Price
  • 470177-094
    Compound Coarse and Fine Focus Beginner Microscope
    Compound Coarse and Fine Focus Beginner Microscope
    Corded Tungsten
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $3,304.64
    $3,304.64
    Each (1UN)
       $3,304.64
    Supplier Number: 5585000
  • 470177-096
    Compound Coarse and Fine Focus Beginner Microscope with Gliding Stage
    Compound Coarse and Fine Focus Beginner Microscope with Gliding Stage
    Cordless LED
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $4,050.85
    $4,050.85
    Each (1UN)
       $4,050.85
    Supplier Number: 5791200