Impact Car
Impact Car
470101-766
: IMPACT1
Add Some Momentum To Your Physics Lessons
- Force indicator notes the maximum force
- Includes Activity Guide
Visually demonstrate the increase in momentum of an object as the mass increases with this sturdy, plastic impact car. Equipped with a graduated spring scale and slide, the car displaces the slide on impact against a wall or similar obstacle. The slide remains in its displaced position until reset, allowing students to take direct readings and measure the force. Allows for repeat experiments with increasing mass. Includes activity guide.
- Description:Impact Car