Imhoff Settling Cone
Imhoff Settling Cone Rack, —
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Supplier #: W990760
Jump to:
Product Details & Documents
Determines Settleable Matter in Water
- Unbreakable polycarbonate construction
- Graduated from 0–1000 mL
This unbreakable, polycarbonate Imhoff cone is perfect for determining the amount of settleable matter in water. It is graduated from 0–1000 mL and has a leak-proof closure for removal of sediment for gravimetric measuring (and for easy cleaning). The aluminum rack holds up to three cones and has a wide base for stability. Size: 18"L x 4" dia.
Documents
Certificates
Search by Lot NumberSpecifications
- Description:Imhoff Settling Cone Rack
- Nominal Capacity:—
- Cat. No.:470205-618