Ileum, Thin Section Slide
Ileum, Thin Section, Thin section, Hematoxylin and Eosin
Catalog # 470183-686
Supplier: Avantor
Jump to:
Product Details & Documents
Extremely Thin Plastic Embedded Tissue For Greater Viewing Clarity
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Shows general structure of ileum.
Especificaciones
- Description:Ileum, Thin Section
- Stain:Hematoxylin and Eosin
- Orientation:Thin section