Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Descripción:Identification of Substances Lab Activity
- No. of Lab Groups:12
- Referencia:470002-484
- Incluye:Teacher Guide, SDS, Student Guide, Worksheets
- Nº de proveedor:83
Specifications
About this item
This lab is a great introduction to qualitative analysis.
- Activities Include Identifying Common Metallic and Non-Metallic Ions in Zinc and Cadmium
- Students Research Distinctive Properties in Tested Substances
- Kit Contains Tests for Known and Unknown Substances
- Safe and Clear Test Results
Specific positive and negative ions are often used to identify inorganic substances. In this kit, students identify common metallic and non-metallic ions in zinc, cadmium, antimony, barium, sulfate, sulfide, sulfite, and carbonate.
Through the identification process, students learn that each substance has distinctive properties. As a closing activity, students conduct chemical tests on four unknown substances. All test results are safe, clear, and colorful.