Orders LinkContactUsLinkComponent
Ward's® Hydroponic Plant Study Lab Activity
WARD’S Hydroponic Plant Study Lab Activity
  470023-820
 :   Avantor
Ward's® Hydroponic Plant Study Lab Activity
  470023-820
 :  Avantor
Ward's® Hydroponic Plant Study Lab Activity
Ward's® Hydroponic Plant Study Lab Activity
WARD’S Hydroponic Plant Study Lab Activity
$3,261.56
Kit de 1 Paleta de 100 (1UN)
Grow plants without soil.
Using hydroponics to grow plants makes it much easier to observe the effects of certain minerals on plant growth. Your students will compare plants grown in soil with plants grown hydroponically. They will also conduct experiments with nutrient solutions lacking key minerals, such as iron and calcium. This will allow them to see first hand how these missing nutrients affect plant growth.
It includes materials for 15 setups, a teacher’s guide, and student copymaster.
  • Description:
    WARD’S Hydroponic Plant Study Lab Activity