General Use Hydrometers
General Use Hydrometers , Type: Wide Range, Ranges: 0.70 - 2.00 x 0.01, Length, cm: 40
Catalog # 470122-484
Supplier: CHASE INSTRUMENTS CO SE
Número de proveedor #: UN2000
Jump to:
Product Details & Documents
Our general use hydrometers are made of smooth glass for easy cleaning and have sealed-in paper scales and metal ballast.
Especificaciones
- Description:General Use Hydrometers , Type: Wide Range, Ranges: 0.70 - 2.00 x 0.01, Length, cm: 40