Orders LinkContactUsLinkComponent
Hydrion Test Paper Strips
Hydrion Test Paper Strips
Certificates
Hydrion Test Paper Strips
Hydrion Test Paper Strips
2 Options Available Below
Product Details & Documents
Use Hydrion A to test in the range of 2-4-6-8-10 and Hydrion B for the range of 1-3-5-7-9-11. Both strips let you test the pH scale in 2.0 units.
Includes: color chart.
Hydrion 1-12 pH Paper is a general use pH paper that can be used for: 
Classroom demonstrations, and general laboratories.
Product Family Options
Información del producto
  • pH Graduation
    pH Range
    Availability
    Price
  • 470200-342
    ph Paper, Hydrion A Test Strips, 10 Vials (1000 Strips Total)
    ph Paper, Hydrion A Test Strips, 10 Vials (1000 Strips Total)
    2
    2 - 10
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (10UN)
    Case (10UN) - $794.20
    $794.20
    Case (10UN)
       $794.20
    Supplier Number: 165A
  • 470200-344
    pH Paper, Hydrion B Test Strips Vials (1000 Strips Total)
    pH Paper, Hydrion B Test Strips Vials (1000 Strips Total)
    2
    1 - 11
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (10UN)
    Case (10UN) - $794.20
    $794.20
    Case (10UN)
       $794.20
    Supplier Number: 165B