Orders LinkContactUsLinkComponent
Hydrion pH Test Papers
Hydrion pH Test Papers
Hydrion pH Test Papers
Certificates
Hydrion pH Test Papers
Hydrion pH Test Papers
4 Options Available Below
Product Details & Documents
Includes Color Comparison Chart.
  • Wide range shows sharp color change for each full ph unit
  • Narrow range papers show a color change for each half pH unit
The wide range (1.0 - 12.0 and 1.0 - 14.0) shows a sharp color change for each full pH unit. The narrow range papers show a color change for each half pH unit. Packages of five rolls.
Product Family Options
Product Information
  • pH Range
    Availability
    Price
  • 470203-378
    pH paper roll
    pH paper roll
    3.0 - 5.5
    Log in for availability
    Log in for availability
    Log in for availability
    Paquete de 5
    Paquete de 5 - $800.26
    $800.26
    Log in for availability
    Cartón de 10 (5UN)
    Cartón de 10 (5UN) - $872.28
    $872.28
    Paquete de 5
       $800.26
    Cartón de 10 (5UN)
       $872.28
    Supplier Number: 425
  • 470203-380
    pH paper roll
    pH paper roll
    6.0 - 8.0
    Log in for availability
    Log in for availability
    Log in for availability
    Paquete de 5
    Paquete de 5 - $800.26
    $800.26
    Log in for availability
    Cartón de 10 (5UN)
    Cartón de 10 (5UN) - $872.28
    $872.28
    Paquete de 5
       $800.26
    Cartón de 10 (5UN)
       $872.28
    Supplier Number: 445
  • pH paper roll
    1.0 - 12.0
    Log in for availability
    Log in for availability
    Log in for availability
    Each (5UN)
    Each (5UN) - $800.26
    $800.26
    Log in for availability
    Case de 10 (5UN)
    Case de 10 (5UN) - $872.28
    $872.28
    Each (5UN)
       $800.26
    Case de 10 (5UN)
       $872.28
    Supplier Number: 160
  • 470175-230
    pH paper roll Insta-Check
    pH paper roll Insta-Check
    1.0 - 14.0
    Log in for availability
    Log in for availability
    Log in for availability
    Paquete de 5
    Paquete de 5 - $891.62
    $891.62
    Log in for availability
    Case de 10 (5UN)
    Case de 10 (5UN) - $971.87
    $971.87
    Paquete de 5
       $891.62
    Case de 10 (5UN)
       $971.87
    Supplier Number: 494