Orders LinkContactUsLinkComponent
Lab-Aids® Human Genetics Experiment Kit
Lab-Aids® Human Genetics Experiment Kit
Human Genetics Experiment Kit, 50
Catalog # 470111-670
Número de proveedor #:  7
Lab-Aids® Human Genetics Experiment Kit
Catalog # 470111-670
Lab-Aids® Human Genetics Experiment Kit
Lab-Aids® Human Genetics Experiment Kit
Human Genetics Experiment Kit, 50
Price
$1,870.85
Log in Para disponibilidad
Kit de 1
Product Details & Documents
How are inherited traits transmitted?
  • Develop an experiment
  • Understand and demonstrate principles of heredity
  • Time required: one 50 minutes class period
The experiment is based on the transmission of a single inherited trait - the ability to taste harmless PTC (phenylthiocarbamide). The goal is for students to determine if they have this gene. They collect data by testing family members, correlating and pooling the data for the entire class, then calculating the random segregation data according to the Hardy-Weinberg Law. Background information about the Hardy-Weinberg Law and information about calculating gene frequency and the genetics of ABO blood typing are included.
Included:
Teacher's Guide with SDS
PTC test paper, strips
Control test paper, strips
Genealogy and data charts
 : See datasheet for contents information.
Especificaciones
  • Description:
    Human Genetics Experiment Kit
  • No. of Lab Groups:
    50