Human Evolution Poster
Evolution Poster, 92 cm, 61 cm
Catalog # 470105-968
Supplier: FEENIXX PUBLISHING, INC.
: A103L
Jump to:
Product Details & Documents
Covers human evolution from primitive humans who walked the Earth over 4 million years ago to anatomically modern humans who existed until about 30, 000 years ago.
- Distinctive, detailed teaching tool
- Laminated finish
Features illustrations of how humans lived throughout the eons, the tools and devices they used, position of the continents and human’s evolving geographic distribution and more.
Especificaciones
- Description:Evolution Poster
- Length:92 cm
- Width:61 cm