Eisco® Circulatory System
Circulatory System, 1, 20 cm, 45 cm, 6 cm
Catalog # 470111-072
Supplier: VWR International
Jump to:
Product Details & Documents
A basic illustration of the circulatory system.
- Mounted on board
- Hand painted
- Includes key card
Gain a general perspective of human blood circulation with this colorful model. It portrays venous blood and arterial blood to give reinforcement to the oxygenation and deoxygenation of hemoglobin as it travels the body’s vascular network. Model is numbered and mounted on baseboard.
Especificaciones
- Description:Circulatory System
- Height:6 cm
- Length:45 cm
- Width:20 cm
- No. of Parts:1