Human Blood Slide
Human Blood, Smear, Giemsa
Catalog # 470177-634
Supplier: Avantor
Jump to:
Product Details & Documents
Investigate the components within your blood
- Identifying features clearly distinguishable
- Rigorous quality control standards
Human. Shows the three major functional classes: red cells, white cells and platelets. Human red blood cells have no nuclei.
Especificaciones
- Description:Human Blood
- Stain:Giemsa
- Orientation:Smear