Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Descripción:How Clean is the Water We Drink and the Air We Breathe? Kit
- No. of Lab Groups:10
- Referencia:470012-370
- Nº de proveedor:S-30
Make the invisible visible. A safe and simple way to teach pollution and contamination, this kit allows students to detect microscopic microbes in samples of air and water they collect.
- Collect local air and water samples
- Observe microorganisms
- Requires a 400× microscope
Simply incubate the samples overnight, then view the following day with a 400X microscope you provide. Kit includes materials for 10 student groups. Kit requires a microscope with 400× to view the incubated samples.