Hook's Law Apparatus
Hook's Law Apparatus, Student, 30 x 12 x 12 cm, 0.5 kg (1.2 lbs.)
Catalog # 470016-050
Supplier: VWR International
Jump to:
Product Details & Documents
This sturdy Hooke's Law apparatus features a mirrored scale to eliminate parallax error.
- Mirrored Scale
- Easy-To-Read Pointer
- Adjustable Scale
- Support Rod
- Wooded Base
Mounted on a 30 cm support rod and wooden base, a hook supporting a coiled spring includes a mass hanger and indicator.
The apparatus is marked in clear millimeters, so students can easily obtain measurements and record data using this device. The apparatus requires weights, available separately.
Specifications
- Description:Hook's Law Apparatus, Student
- L×W×H:30 x 12 x 12 cm
- Weight:0.5 kg (1.2 lbs.)