Orders LinkContactUsLinkComponent
Hook Collars
Hook Collars
Hook Collars
Hook Collars
Quickly add a hook to a support rod.
  • Plated steel
  • Thumbscrew
Plated steel collar clamps feature thumbscrews and hooks for suspending pulleys, balances, etc. from support rods or crossbars.
Product Family Options
  • Description
  • Hook Collar, Fits 13 mm (0.5") Rod
    Each (1UN)
    Each (1UN) - $103.94
    $103.94
    Each (1UN)
       $103.94
     : HKCL01
  • Hook Collar, Fits 19 mm (0.75") Rod
    Each (1UN)
    Each (1UN) - $87.42
    $87.42
    Each (1UN)
       $87.42
     : HKCL02