Hollow Prism
Hollow Prism
Catalog # 470012-844
Supplier: UNITED SCIENTIFIC SUPPLIES
Número de proveedor #: PHA045
Jump to:
Product Details & Documents
Perform Several Optics Experiments
- This unique optics kit comes with a clear hollow prism, funnel, and plug
- Measures 2 1/2" H x 1 7/8" W x 1 13/16" D
Young scientists can fill it with clear or colored liquid to explore light refraction and the visible light spectrum. A perfect addition to the classroom for demonstrations or science projects.
Documents
Hollow Acrylic Prism
Videos
Especificaciones
- Description:Hollow Prism