Hoffman Screw-Compressor Clamps for Flexible Tubing
Catalog # 470005-380
Supplier: UNITED SCIENTIFIC SUPPLIES
Specifications
- Descripción:Hoffman Screw-Compressor Clamps for Flexible Tubing, Open Jaw
- Referencia:470005-380
- Nº de proveedor:HSCOP2
Specifications
About this item
Easily clamp tubes up to 1" in diameter.
- Zinc-plated brass
Can use as a closed clamp or open.