Orders LinkContactUsLinkComponent
High-Frequency Tesla Coil
High-Frequency Tesla Coil
High-Frequency Tesla Coil
Certificates
High-Frequency Tesla Coil
High-Frequency Tesla Coil
1 Options Available Below
Product Details & Documents
The high-frequency coil can be used to test vacuum equipment for leakage or excite electrical tubes for demonstrations.
  • Coil tests vacuum equipment for leakage or excites electrical tubes for demonstrations
  • Instructions included
  • Connecting cord and attachment plug included
  • Coil is mounted in Bakelite case
  • Small enough to hold in one hand
It's mounted in a hard Bakelite case small enough to hold in one hand. One terminal is attached to the pointed end of the coil. A screw at the large end is adjustable.
The voltage is 40 to 50 kV, and the spark frequency is three to four MHz. A featured knob adjusts the spark length. After 10 minutes of continuous use, the coil must be turned off to cool.
Ordering information: This coil includes a connecting cord, attachment plug, and instructions.
Product Family Options
Información del producto
  • Availability
    Price
  • 470005-396
    High-frequency coil, 110 V
    High-frequency coil, 110 V
    Discontinued
    Supplier Number: -
  • 470122-120
    High-frequency coil, with switch
    High-frequency coil, with switch
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $14,604.39
    $14,604.39
    Each (1UN)
       $14,604.39
    Supplier Number: 11031