Orders LinkContactUsLinkComponent
Ward's® Hematite (Red Ochre)
Ward's® Hematite (Red Ochre)
Supplier:   Avantor
Ward's® Hematite (Red Ochre)
Supplier:  Avantor
Certificates
Ward's® Hematite (Red Ochre)
Ward's® Hematite (Red Ochre)
3 Options Available Below
Product Details & Documents
Red, earthy, in natural chunk form; common pigment ore.
  • Collected in South Dakota
  • Multiple sizes and pack options to fit your specific needs
  • Cut by Ward's geologists to guarantee top market quality
Student Specimens – Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand Specimens – Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk Pack – Includes 1 kilo of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25" to 2×3". Packaged in a labeled storage box for easy organization.
All specimen dimensions are considered an average size range.
Product Family Options
Specimens
Información del producto
  • Specimen Dimensions
    Availability
    Price
  • 470025-454
    Hematite (Red Ochre), student specimens
    Hematite (Red Ochre), student specimens
    1×1 - 1×1"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 10
    Paquete de 10 - $278.35
    $278.35
    Paquete de 10
       $278.35
  • 470025-458
    Hematite (Red Ochre), hand specimen
    Hematite (Red Ochre), hand specimen
    2×3" - 3×4"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $299.55
    $299.55
    Each (1UN)
       $299.55
  • 470025-456
    Hematite (Red Ochre), bulk pack, 1 kg
    Hematite (Red Ochre), bulk pack, 1 kg
    Size will vary
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $430.70
    $430.70
    Each (1UN)
       $430.70