Orders LinkContactUsLinkComponent
Ward's® Hematite (Oolitic)
Ward's® Hematite (Oolitic)
 :   Avantor
Ward's® Hematite (Oolitic)
 :  Avantor
Ward's® Hematite (Oolitic)
Ward's® Hematite (Oolitic)
Red, classic oolitic texture.
  • Collected in New York
  • Multiple sizes and pack options to fit your specific needs
  • Cut by Ward's geologists to guarantee top market quality
  • Matching thin section available
Student Specimens – Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand Specimens – Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk Pack – Includes 1 kilo of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25" to 2×3". Packaged in a labeled storage box for easy organization.
All specimen dimensions are considered an average size range.
Product Family Options
Specimens
  • Description
    Specimen Dimensions
  • Hematite (Oolitic), student specimens
    1×1 - 1×1"
    Paquete de 10
    Paquete de 10 - $249.73
    $249.73
    Paquete de 10
       $249.73
  • Hemitate (Oolitic), hand specimen
    2×3" - 3×4"
    Each (1UN)
    Each (1UN) - $250.85
    $250.85
    Each (1UN)
       $250.85
  • Hematite (Oolitic), bulk pack, 1 kg
    Size will vary
    Each (1UN)
    Each (1UN) - $318.45
    $318.45
    Each (1UN)
       $318.45