Heat Conductometer
Heat Conductometer, 38 x 18.4 cm, 0.5 kg (1.1 lbs.)
Catalog # 470148-760
Supplier: VWR International
Jump to:
Product Details & Documents
This unit visually reinforces heat conductivity lessons.
- Five metals included: aluminum, brass, copper, nickel, andsteel
- Instructions included
It demonstrates how five different metals—aluminum, brass, copper, nickel, and steel—conduct heat. The five rods are heated over a flame simultaneously, and students gauge what metal conducts heat best by observing how fast the wax at the tip of each rod melts.
Ordering information: The apparatus comes with instructions. Wax and a heat source are required but not included.
Especificaciones
- Description:Heat Conductometer
- L×W:38 x 18.4 cm
- Weight:0.5 kg (1.1 lbs.)