Hand Centrifuge
Hand Centrifuge
Catalog # 470121-084
Supplier: UNITED SCIENTIFIC SUPPLIES
Supplier #: CENTFG1
Jump to:
Product Details & Documents
This hand-driven centrifuge is ideal for simple classroom demonstrations.
- Easy grip handle
- Holds four tubes at once
The centrifuge features a painted metal body and head that can hold up to four tubes, which are included, and comes with a table-mounting clamp. The three-piece construction makes for easy disassembly and storage in its molded foam box.
Documents
Especificaciones
- Description:Hand Centrifuge