CENCO Hall Effect Apparatus
Hall Effect Apparatus
470121-160
: Avantor
This apparatus determines hall voltage and coefficient.
- Heavy-Duty Electromagnet
- Gauss Meter with a Thin Probe
- Manual
- Semiconductor Ge Sample
- Digital Multimeter
The most distinguishing feature allows the student to individually mount a Ge crystal.
Students can also determine the type of charge carrier, the charge density of carriers, and the resistivity of a given sample.
: The unit includes a Gauss meter with probes; digital multimeter; semiconductor 'Ge' sample, p type; and Hall effect apparatus. A 12V power supply is required but not provided.
- Description:Hall Effect Apparatus