Orders LinkContactUsLinkComponent
Stopcock/Vacuum Grease
150 g
  470146-194
 :   Liveo
 :  1597418
Stopcock/Vacuum Grease
  470146-194
 :  Liveo
Stopcock/Vacuum Grease
Stopcock/Vacuum Grease
150 g
$960.42
Each (1UN) Case de 12 (1UN)
Highly Resistant to Water, Alkalis, and Chemicals
  • Silicone based lubricant for general use
  • Sealant for vacuum and pressure systems
  • Has a low volatility
  • Withstands temperatures from -40 to 260°C
  • Size:
    150 g