Ward's® Gravity and Orbits Lab Activity
Catalog # 470015-222
Supplier: Avantor
Specifications
- Descripción:Ward's® Gravity and Orbits Lab Activity
- No. of Lab Groups:5
- Referencia:470015-222
- Nº de proveedor:WARD470015-222
Specifications
About this item
Enough Materials to Construct Five Sets of Two Models
- Includes a teacher's guide
Students investigate the effect of gravity on a moving body, then determine the relationship between a celestial object’s period of revolution and the force pulling on the object, by constructing two simple models in a two-part lab activity. You will receive enough materials for five setups, a teacher’s guide, and student copymasters.