Orders LinkContactUsLinkComponent
Ward's® Granite (Pink)
Ward's® Granite (Pink)
 :   Avantor
Ward's® Granite (Pink)
 :  Avantor
Ward's® Granite (Pink)
Ward's® Granite (Pink)
Medium-grained with biotite.
  • Collected in Rhode Island
  • Multiple sizes and pack options to fit your specific needs
  • Cut by Ward's geologists to guarantee top market quality
  • Matching thin section available
Student specimens - Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand specimens - Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk pack - Includes 1 kilo of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25" to 2×3". Packaged in a labeled storage box for easy organization.
All specimen dimensions are considered an average size range.
Product Family Options
Specimens
  • Description
    Specimen Dimensions
  • Granite (Pink), Student Specimens
    1×1 - 1×1"
    Paquete de 10
    Paquete de 10 - $222.80
    $222.80
    Paquete de 10
       $222.80
  • Granite (Pink), Hand Specimen
    2×3" - 3×4"
    Each (1UN)
    Each (1UN) - $240.55
    $240.55
    Each (1UN)
       $240.55
  • Granite (Pink), Bulk Pack, 1 kg
    Size will vary
    Each (1UN)
    Each (1UN) - $321.30
    $321.30
    Each (1UN)
       $321.30