Polypropylene Cylinders with Handle
Supplier: BEL-ART PRODUCTS SE
Acerca de este artículo
Put an End to the Difficult Task of Pouring from Large, Heavy Cylinders.
- Double scale raised graduations
- Chemically resistant
- Autoclavable to 121°C
2 Options Available Below
Product Details & Documents
Put an End to the Difficult Task of Pouring from Large, Heavy Cylinders.
- Double scale raised graduations
- Chemically resistant
- Autoclavable to 121°C
Specifications for Product Family
Especificaciones
- Catalog No:
- 470012-892
- 470003-612
- Form:
- Tall
- Tall
Especificaciones
Product Family Options
Información del producto
- CapacityGraduation IntervalGraduation RangeAvailabilityPrice
- Supplier Number: F28461-1000_SEEspecificaciones:
More
Specifications
Cat. No.470012-892FormTall - Supplier Number: F28461-2000_SEEspecificaciones:
More
Specifications
Cat. No.470003-612FormTall