Gold Foil Electroscope
Gold foil electroscope
470020-658
: ELGL01
A Highly Sensitive Electroscope
- Highly sensitive gold foil
- Detect small static charges
This high sensitive electroscope can detect charges that a regular electroscope cannot. A gold leaf is mounted on an insulated rod in a grounded metal case, and the glass windows at the front and back of the case allow for easy observation or projection. It is a good alternative for high humidity environments.
- Description:Gold foil electroscope
- Cat. No.:470020-658