ContactUsLinkComponent
GLOBE® Soil Color Book
GLOBE® Soil Color Book
Catalog # 470005-702
Supplier:  PECO SALES INC SE
undefined
GLOBE® Soil Color Book
Catalog # 470005-702
Supplier:  PECO SALES INC SE
Supplier Number:  77369
Specifications
  • Descripción:
    GLOBE Soil Color Book
  • Referencia:
    470005-702
  • Nº de proveedor:
    77369
Specifications
About this item
Created especially to meet the requirements of the GLOBE program.
  • Aligned with GLOBE protocols
The color chips in this spiral-bound book are specially formulated using more earth-based pigments to make it easier to match under less than perfect lighting conditions. Over 200 chips and a soil structure key are included in the set. Size: 7 1/4" x 3 1/4".